Anti-FUT4 Mouse Monoclonal Antibody [clone: FR4A5]

Supplier: Biotium

BNUB0504-100 BNUB0504-500
75888-032EA 240.92 USD
75888-032 75888-034
Anti-FUT4 Mouse Monoclonal Antibody [clone: FR4A5]
Antibodies
CD15 plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on  more than 95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. In addition, CD15 is expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin’s disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

Note: Conjugates of blue fluorescent dyes like CF®405S and CF®405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: FUT4
Clonality: Monoclonal
Clone: Clone FR4A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR