Anti-ODC1 Mouse Monoclonal Antibody (Biotin) [clone: ODC1/486]

Supplier: Biotium

BNCB0486-100 BNCB0486-500
75966-680EA 205.77 USD
75966-680 75966-682
Anti-ODC1 Mouse Monoclonal Antibody (Biotin) [clone: ODC1/486]
Antibodies
This antibody recognizes a 53 kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa.

Type: Primary
Antigen: ODC1
Clonality: Monoclonal
Clone: Clone ODC1/486
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR