Anti-CDw60 Mouse Monoclonal Antibody [clone: UM4D4]

Supplier: Biotium

BNUB0481-100 BNUB0481-500
75909-424EA 240.92 USD
75909-424 75909-426
Anti-CDw60 Mouse Monoclonal Antibody [clone: UM4D4]
Antibodies
This MAb reacts with 9-O-acetylated disialosyl group linked to the glycoprotein CD60w. CDw60 is a carbohydrate antigen present on gangliosides, platelets and a subset of CD4 , CD8 peripheral T-cells, but not on B-cells, monocytes or granulocytes. CDw60 is involved in the co-stimulation of T-cells.

Type: Primary
Antigen: CDw60
Clonality: Monoclonal
Clone: Clone UM4D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR