Anti-SILV Mouse Monoclonal Antibody (CF488A) [clone: HMB45]

Supplier: Biotium

BNC880444-500 BNC880444-100
75970-290EA 569.73 USD
75970-290 75970-896
Anti-SILV Mouse Monoclonal Antibody (CF488A) [clone: HMB45]
Antibodies
By immunohistochemistry, it specifically recognizes a protein in melanocytes and melanomas. This MAb reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin. Metastatic amelanotic melanoma can often be confused with a variety of poorly differentiated carcinomas, large cell lymphomas, and sarcomas using H & E stains alone. It is also difficult to differentiate melanoma from spindle cellMore Product Information
Order Now

SPECIFICATIONS

Antigen Symbol SILV
Environmentally Preferable
Antigen Name Pmel17 / gp100 / SILV
Antibody Type Primary
Clonality Monoclonal
Conjugation CF® 488A
Clone HMB45
Reactivity Human
Host Mouse
Gene ID Unigene ID: 95972
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage Buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR