Anti-Biotin Mouse Monoclonal Antibody (CF405S) [clone: Hyb8]

Supplier: Biotium

BNC040400-100 BNC040400-500
75885-322EA 205.77 USD
75885-322 75885-324
Anti-Biotin Mouse Monoclonal Antibody (CF405S) [clone: Hyb8]
Antibodies
This antibody recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. This MAb is highly specific to biotin and shows no cross-reaction with other structurally related compounds. It has a very high affinity for biotin and is excellent for use in various amplification techniques. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: Clone Hyb8
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR