Anti-TACSTD1 Mouse Monoclonal Antibody (CF640R) [clone: 323/A3]

Supplier: Biotium

BNC400396-100 BNC400396-500
75893-266EA 205.77 USD
75893-266 75893-268
Anti-TACSTD1 Mouse Monoclonal Antibody (CF640R) [clone: 323/A3]
Antibodies
EGP40 is a 40-43 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. This antibody has been used to distinguish adenocarcinoma from pleural mesothelioma and hepatocellular carcinoma. This antibody is also useful in distinguishing serous carcinomas of the ovary from mesothelioma.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: TACSTD1
Clonality: Monoclonal
Clone: Clone 323/A3
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR