Anti-MS4A1 Mouse Monoclonal Antibody (CF488A) [clone: 109-3C2]

Supplier: Biotium

BNC880355-100 BNC880355-500
75890-486EA 205.77 USD
75890-486 75890-488
Anti-MS4A1 Mouse Monoclonal Antibody (CF488A) [clone: 109-3C2]
Antibodies
This antibody recognizes a protein of 30-33 kDa, which is identified as CD20 (Workshop V; Code CD20.12. Workshop IV; Code B17). It This antibody recognizes an extracellular domain of CD20. It is a non-Ig differentiation antigen of B-cells and its expression is restricted to normal and neoplastic B-cells, being absent from all other leukocytes and tissues. CD20 is expressed by pre B-cells and persists during all stages of B-cell maturation but is lost upon terminal differentiation into plasma cells. The protein passes through the membrane 4 times with both ends in cytoplasm and exposes one short and one longer loop to the external environment. CD20 is not glycosylated in resting B-cells and its cytoplasmic domains are differentially phosphorylated upon activation. It acts as calcium channel involved in B cell activation and cell cycle progression.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®488A is a green fluorescent dye (Ex/Em 490/515 nm) with excellent brightness and photostability. The dye is minimally charged for less non-specific binding. CF®488A also is compatible with super-resolution imaging by TIRF.

Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: Clone 109-3C2
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG3, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR