Anti-CD3E Mouse Monoclonal Antibody (Biotin) [clone: RIV9]

Supplier: Biotium

BNCB0322-100 BNCB0322-500
75895-266EA 205.77 USD
75895-266 75895-268
Anti-CD3E Mouse Monoclonal Antibody (Biotin) [clone: RIV9]
Antibodies
This antibody reacts with five invariable CD3 chains designated CD3y, CD3d, CD3e, CD3f, and CD3n with molecular weight ranging from 16-28 kDa. It is found on all mature T-lymphocytes, NK, T-cells, and some thymocytes. CD3, also known as T3, is a member of the immunoglobulin superfamily that plays a role in antigen recognition, signal transduction, and T cell activation. It is expressed, typically at high levels, on peripheral T cells and majority of T cell neoplasms. Thymocytes express CD3 at different level on the cell surface in the course of differentiation and, in cortical thymus, CD3 is predominantly intracytoplasmic. This MAb is particularly useful for induction of T cell activation in vitro and in vivo due to its unusual IgG3 isotype.

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: Clone RIV9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG3, kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR