Anti-TFRC Mouse Monoclonal Antibody [clone: DF1513]

Supplier: Biotium

BNUB0188-100 BNUB0188-500
75918-044EA 240.92 USD
75918-044 75918-046
Anti-TFRC Mouse Monoclonal Antibody [clone: DF1513]
Antibodies
This antibody recognizes a ~90-95 kDa protein which is identified as cell surface transferrin receptor (CD71), a disulfide-bonded homodimeric glycoprotein of 180-190 kDa (Workshop IV). This MAb is highly specific to CD71 and shows no cross-reaction with other related proteins. Ligand for transferrin receptor is the serum iron transport protein, transferrin. This receptor is broadly distributed in carcinomas, sarcomas, leukemias, and lymphomas. CD71/Transferrin receptor has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinical behavior or response to therapy in a number of malignancies including breast cancer.

Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: Clone DF1513
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR