Anti-ICAM3 Mouse Monoclonal Antibody (CF488A) [clone: 186-2G9]

Supplier: Biotium

BNC880178-100 BNC880178-500
75915-052EA 205.77 USD
75915-052 75915-054
Anti-ICAM3 Mouse Monoclonal Antibody (CF488A) [clone: 186-2G9]
Antibodies
This antibody recognizes the D1 domain of an N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3 (Leucocyte Workshop VI). CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This MAb is excellent for staining of formalin/paraffin tissues.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®488A is a green fluorescent dye (Ex/Em 490/515 nm) with excellent brightness and photostability. The dye is minimally charged for less non-specific binding. CF®488A also is compatible with super-resolution imaging by TIRF.

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: Clone 186-2G9
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR