Anti-ITGAL Mouse Monoclonal Antibody (CF543) [clone: Cris-3]

Supplier: Biotium

BNC430151-100 BNC430151-500
75886-378EA 205.77 USD
75886-378 75886-556
Anti-ITGAL Mouse Monoclonal Antibody (CF543) [clone: Cris-3]
Antibodies
This antibody recognizes a protein of 180 kDa, identified as CD11a (Leucocyte Workshop II; Code N202). CD11a complex with the 2 subunit of the integrin family, CD18, to form the cell surface heterodimer, LFA-1 or CD11a /C18 (aLbL). LFA-1 is expressed on all leukocytes including lymphocytes, monocytes, and granulocytes. It is involved in leukocyte adhesion to its ligands including intercellular adhesion molecule-1 (ICAM-1 or CD54), ICAM-2 (CD102), ICAM-3 (CD50) and Telencephalin (TLN) and play a role in most immune/inflammatory responses.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®543 is an orange fluorescent dye (Ex/Em 541/560 nm) designed for the 543 nm laser line. It yields the brightest conjugates among spectrally similar dyes.

Type: Primary
Antigen: ITGAL
Clonality: Monoclonal
Clone: Clone Cris-3
Conjugation: CF543
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR