Anti-UCHL1 Mouse Monoclonal Antibody [clone: 13C4]

Supplier: Biotium

BNUB0147-100 BNUB0147-500
75969-710EA 240.92 USD
75969-710 75969-712
Anti-UCHL1 Mouse Monoclonal Antibody [clone: 13C4]
Antibodies
This MAb reacts with a protein of 20-30 kDa, identified as PGP 9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UCHL-1). Initially, PGP 9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP 9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP 9.5 gene is believed to cause a form of Parkinson's disease.

Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: Clone 13C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR