DuPont™ ProShield® 50 Coveralls with Elastic Wrists and Ankles
Supplier: DuPont
NB125SWH2X002500 NB125SWH3X002500 NB125SWH4X002500 NB125SWH5X002500 NB125SWH6X002500 NB125SWHLG002500 NB125SWHMD002500 NB125SWHSM002500 NB125SWHXL002500
76187-528CS 124.33 USD
76187-528 76187-530 76187-532 76187-534 76187-536 76187-538 76187-540 76187-542 76187-544
DuPont™ ProShield® 50 Coveralls with Elastic Wrists and Ankles
Disposable Garments
ProShield® 50 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Collared
  • Elastic wrists and ankles
  • Serged seams
ProShield® 50 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include: janitorial, sanitation, and general industrial maintenance.
Certificaciones: Thickness meets ASTM D751.