DuPont™ Tychem® 5000 Coveralls with Standard Hood and Attached Socks
Supplier: DuPont
C3122TTNXL000600 C3122TTN2X000600 C3122TTN3X000600 C3122TTN4X000600 C3122TTN5X000600 C3122TTN6X000600 C3122TTN7X000600 C3122TTNLG000600 C3122TTNMD000600 C3122TTNSM000600
76187-242CS 540.68 USD
76187-242 76187-244 76187-690 76187-692 76187-694 76187-696 76187-698 76187-700 76187-702 76187-704 76187-706 76187-708 76187-710 76187-712 76187-714 76187-716 76187-718 76187-720
DuPont™ Tychem® 5000 Coveralls with Standard Hood and Attached Socks
Disposable Garments
Tychem® 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate.
  • Standard fit hood
  • Elastic wrists
  • Attached socks
  • Storm flap with adhesive closure
  • Taped seams
Tychem® 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist. These coveralls are tan.
Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Attached hood with elastic around face opening is a standard fit. Strong zipper ensures a tough, flexible closure. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Elastic opening at wrists allows for tighter fit. Integrated socks are composed of the same garment material.
Certificaciones: Thickness meets ASTM D1777.