HYPO-CHLOR® Sodium Hypochlorite Neutral, Veltek

Supplier: Veltek Associates
SHC-NPH-0.25-02 SHC-NPH-0.52-16Z SHC-NPH-0.52-02 SHC-NPH-0.25-16Z
76102-184CS 361.7 USD
76102-184 76102-186 76102-188 76102-190
HYPO-CHLOR® Sodium Hypochlorite Neutral, Veltek
Cleaning Agents Disinfectants
HYPO-CHLOR® Neutral Products are a ready-to-use and neutralized 0.52% and 0.25% sodium hypochlorite solutions formulated with Water for Injection (WFI).

  • Filled in an ISO 5 (Grade A/B, Former Class 100)
  • Filtered at 0.2 microns
  • Components are air washed with 0.2 micron filtered air
  • Aseptically filled into sterile components via gamma irradiation
  • Lot sterility tested according to current USP compendium
  • Completely traceable from start to finish
  • Safe to use daily on stainless steel surfaces

Designed for all pharmaceutical, biopharmaceutical, biotechnology, health care, medical device, and diagnostic manufacturing cleaning rotations that demand a neutral pH and sterile sodium hypochlorite solution adequate for maintaining a clean and critical environment. Compatible with most hard, non-porous, environmental surfaces and while reducing the consequences of corroding, rusting, or pitting.

Applications include: Cleanrooms and controlled areas with surfaces which are hard, non-porous, inanimate surfaces in aseptic filling, gowning rooms, general manufacturing areas, and laboratories. Also on machinery, tables, counters, laminar flow benches, floors, walls, carts, tools, shelves, made of plastic, glass, vinyl, glazed porcelain, laminates, glazed tiles, and stainless steel.

Certificaciones: Delivered with lot specific Certificate of Analysis and Certificate of Sterility.

Envase: Double bagged in easy tear bag.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR