VECTASTAIN® ABC-Alkaline Phosphatase Kit, Vector Laboratories
Supplier: Vector Laboratories
AK-5002 AK-5200 AK-5000 AK-5001 AK-5004
101097-534EA 718.62 USD
101097-534 101097-550 101097-530 101097-532 101097-538
VECTASTAIN® ABC-Alkaline Phosphatase Kit, Vector Laboratories
Blotting Kits
VECTASTAIN® ABC kits take advantage of the flexibility and stability of avidin/biotin binding for sensitive, cost-effective chromogenic target protein detection on tissue, cells, and blots.
  • Avidin/biotin complex (ABC) kits offer a high level of sensitivity and flexibility
  • The VECTASTAIN® ABC-AP Kit (Standard) may be used to detect any biotinylated target, not just secondary antibodies
  • Relatively low-cost protein detection
With more than 50000 citations, Vector Laboratories ABC kits are trusted by immunohistochemistry (IHC) and immunocytochemistry (ICC) laboratories to provide reliable results. The system is modular and different kit variations can accommodate a wide array of primary antibody and tissue species. The kits use alkaline phosphatase (AP) as a detection enzyme. Several kits also use a secondary antibody to target primary antibodies. The ABC complex, once formed, is stable and can be used for several days after preparation.