TAT Broth Base, HiMedia Laboratories
Supplier: HiMedia
GM562-2.5KG GM562-500G GM562-5KG
75804-298EA 772.76 USD
75804-298 75804-300 75804-302
TAT Broth Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
TAT Broth base, granulated with added polysorbate 20 is used for sterility testing of highly viscous or gelatinous substances such as salves, ointments and other cosmetic products.
  • Off-white to yellow colored
  • Granular medium
  • Reaction of 2.5% w/v aqueous solution containing 4% v/v polysorbate 20 at 25 °C
Precaución: Store below 30 °C in tightly closed container and prepared medium at 2 to 8 °C.