HiVeg™ Tributyrin Agar Base, HiMedia Laboratories
Supplier: HiMedia
MV157-500G MV157-2.5KG MV157-5KG
61000-694EA 217.18 USD
61000-694 89228-112 89228-114
HiVeg™ Tributyrin Agar Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Used for the detection of lipolytic microorganisms.
  • Light yellow colored
  • Opalescent gel forms, with oil droplets in petri plates
  • Reaction of 2.3% w/v aqueous solution containing 1% v/v tributyrin at 25 °C
Precaución: Store the dry medium below 30 °C and the prepared medium at 2 to 8 °C.