Plate Stand and Clamp, Promega
Supplier: Promega Corporation
V8261 V8251
PAV8261EA 285.63 USD
PAV8261 PAV8251
Plate Stand and Clamp, Promega
Well Plate Stands and Well Orienters
The Plate Clamp 96 is designed to ensure that nonskirted PCR plates are uniformly flat for liquid transfer on a robotic platform. Recommended for automated protocols. Not designed for skirted PCR plates.
The Plate Stand positions the Plate Clamp 96 (V8251) assembly on the deck of a robotic workstation.
Información para pedidos: Plate Clamp and Plate Stand have to be ordered separately.