Human Cot DNA, Enzo Life Sciences
Supplier: Enzo Life Sciences
GEN116-0500
76202-436EA 314 USD
76202-436
Human Cot DNA, Enzo Life Sciences
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
Blocks repetitive sequences to prevent nonspecific binding.
  • Validated for use in aCGH microarrays and FISH assays
  • Decreases background noise to improve signal fidelity
  • Prevents nonspecific binding and cross-hybridization in samples
Human Cot DNA blocks repetitive sequences of DNA to suppress the level of cross-hybridization as well as non-specific binding to array surfaces. Utilizing Human Cot DNA in hybridizations increases overall sensitivity and improves experimental results.