Albumin (Mouse) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
ALB11-K01
ALB11-K08
ALB11-K04
102993-900EA
1405.77
USD
102993-900
102993-902
103389-042
Albumin (Mouse) ELISA, Eagle Biosciences
Assays
ELISAs
Mouse Albumin ELISA is to detect albumin in mouse urine samples.
- Dynamic Range: 2.5 to 10240 ng/ml
- Sensitivity: 0.1 µg/ml
- Incubation: 2 hours
This kit is designed for the quantitative determination of albumin in mouse urine samples.
Precaución: For research use only and should not be used in diagnostic procedures.