ESF Delta Series, Cell Culture Medium, Expression Systems
Supplier: Expression Systems
96-200 96-299
100000-014EA 127.69 USD
100000-014 103194-132
ESF Delta Series, Cell Culture Medium, Expression Systems
Cell Culture Media
ESF 921 Delta Series formulations are based on the popular ESF 921 medium and are available in a variety of amino acid deficient formats used for structural analysis and protein labeling
Off the shelf formulations include all amino acid deficient and methionine deficient.
Información de suministro: Methionine deficient: Standard Ground.
Custom Deficient: 20L minimum order and are delivered in approximately 6 weeks.