Xanthine Oxidase (from Buttermilk)
Supplier: MilliporeSigma
682151-50U
682151-10U
80057-716EA
325.85
USD
80057-716
80057-714
Xanthine Oxidase (from Buttermilk)
Enzymes
Native xanthine oxidase from buttermilk
Complex metalloprotein that catalyzes oxidative hydroxylation of a variety of aromatic heterocycles and simple aldehydes. Catalyzes the oxidation of xanthine to uric acid.