Ammonium persulfate (APS) 98+% ACS, Ultrapure
Supplier: Thermo Fisher Scientific
|
Danger
|
J76322-22
J76322-A1
J76322-09
101174-878EA
45.2
USD
101174-878
101175-856
101175-858
Ammonium persulfate (APS) 98+% ACS, Ultrapure
Ammonium peroxodisulphate (APS)
Formula:
(NH₄)₂S₂O₈ Melting Pt: 120 °C (dec.) Density: 1.982 Storage Temperature: Ambient |
MDL Number:
MFCD00003390 CAS Number: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1,III Merck Index: 14,541 |
Specification Test Results
Assay (unspecified) | ≥98.0% |
Assay (Aqueous Acidbase Titration) - Titratable Free Acid | ≤0.04 mEq/g |
Chloride Content Chloride and Chlorate (Cl) | ≤0.001% |
Residue on Ignition | ≤0.05% |
Insoluble Material | ≤0.005% |
Heavy Metals (Pb) | ≤0.005% |
Iron (Fe) | ≤0.001% |
Manganese (Mn) | ≤0.5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...