ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form) ≥93%

Supplier: Thermo Fisher Scientific

Synonyms: NADP monosodium salt, NADP Sodium, Nadide phosphate monosodium salt, Sodium (2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)(hydroxy)phosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl hydrogen phosphate, Nicotinamide-adenine dinucleotide phosphate monosodium salt, NADP sodium salt, NADP-Na

J22655-MD J22655-06 J22655-ME
101173-080EA 196.89 USD
101173-080 101174-730 101170-156 101175-200
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form) ≥93%
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)
Formula: C₂₁H₂₇N₇NaO₁₇P₃
MW: 765.39 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036973
CAS Number: 1184-16-3

Order Now

Specification Test Results

Assay (Enzymatic) ≥93%
Residual Water ≤8.0%
Molar Extinction Coefficient (Molar Absorptivity) epsilon₂₆₀ (pH 7.5) [18.0 ±0.8]×10³
Molar Extinction Coefficient (Molar Absorptivity) epsilon₃₄₀ (pH 7.5) [6.2 ±0.4]×10³
Spectral Ratio A₂₅₀/A₂₆₀ (pH 7.5) 0.83 ±0.03
Spectral Ratio A₂₈₀/A₂₆₀ (pH 7.5) 0.21 ±0.02
Spectral Ratio (pH 7.5, Reduced with G6PDH), A₂₆₀/A₃₄₀ 2.33 ±0.11
Spectral Ratio (pH 7.5, Reduced with G6PDH), A₃₄₀/A₂₆₀ 0.43 ±0.02
Sodium (Na) 3.0 ±1.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR