DL-Serine (not of animal origin) 98.5+% (dry basis)
Supplier: Thermo Fisher Scientific
Synonyms:
(±)-Serine, H-DL-Ser-OH, DL-2-Amino-3-hydroxypropionic acid
J21486-22
J21486-A1
101375-170EA
215.58
USD
101375-170
101375-168
DL-Serine (not of animal origin) 98.5+% (dry basis)
DL-Serine
Formula:
C₃H₇NO₃ MW: 105.09 g/mol Boiling Pt: 395 Melting Pt: ∼ 240 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064223 CAS Number: 302-84-1 EINECS: 206-130-6 Merck Index: 14,8460 |
Specification Test Results
Assay (Dry Basis) | ≥98.5% |
Chloride Content | ≤0.021% |
Identification (FTIR) | Conforms |
Residue on Ignition | ≤0.1% |
Impurity Content Sulphate (SO₄) | ≤0.05% |
Loss on Drying | ≤0.3% |
Heavy Metals (Pb) | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...