D-(+)-Raffinose pentahydrate
Supplier: Thermo Fisher Scientific
Synonyms:
D-(+)-Raffinose pentahydrate, Gossypose pentahydrate, (3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrate, Melitriose pentahydrate, O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose pentahydrate
J21060-36
J21060-A1
J21060-22
101170-082EA
748.46
USD
101170-082
101174-646
101174-648
D-(+)-Raffinose pentahydrate
D-(+)-Raffinose pentahydrate
Formula:
C₁₈H₃₂O₁₆·5H₂O MW: 594.52 g/mol Boiling Pt: 78…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00071590 CAS Number: 17629-30-0 EINECS: 208-146-9 Merck Index: 14,8089 |
Specification Test Results
Identification (FTIR) | Conforms |
Residue on Ignition | ≤0.05% |
Loss on Drying | ≤15.3% |
Optical Rotation (c=4 in H₂O and NH₃) | +104.9 ±1.3° |
Arsenic (As) | ≤0.5 ppm |
Heavy Metal (Cu) | ≤10 ppm |
Iron (Fe) | ≤5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...