Polyethylene glycol 6,000, flakes, Ultrapure

Supplier: Thermo Fisher Scientific

Synonyms: PEG 6000PEG 6,000

J19972-A7 J19972-A1
101170-018EA 423.23 USD
101170-018 101175-668
Polyethylene glycol 6,000, flakes, Ultrapure
Polyethylene glycol 6,000

Application includes differential precipitation of DNA, enhances hybridisation rate of nucleic acids, improves efficiency of end-labeling with T4 polynucleotide kinase, used in ligation of blunt ended DNA, precipitates bacteriophage ± particles and plasmid DNA, used in preparing single-stranded M13 DNA and promotes cell fusion.

Formula: H(OCH₂CH₂)nOH
MW: 6000 g/mol
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD01779614
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,7568

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR