L(+)-Glutamic acid 98.5-101.5% (dry basis), Ultrapure
Supplier: Thermo Fisher Scientific
Synonyms:
(S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
J16215-A1
J16215-A7
J16215-C4
101170-914EA
98.32
USD
101170-914
101169-890
101172-550
L(+)-Glutamic acid 98.5-101.5% (dry basis), Ultrapure
L(+)-Glutamic acid
Formula:
HO₂CCH₂CH₂CH(NH₂)CO₂H MW: 147.13 g/mol Boiling Pt: 325 Melting Pt: 189…192 °C (dec.) Density: 1.525 Storage Temperature: Ambient |
MDL Number:
MFCD00002634 CAS Number: 56-86-0 EINECS: 200-293-7 Merck Index: 14,4469 |
|
Specification Test Results
Assay (Unspecified) - Dry Basis | 98.5 - 101.5% |
Identification (FTIR) | Conforms |
Loss on Drying | ≤0.1% |
Optical Rotation (c=10 in 2 N HCl) | 31.2° - 32.2° |
Arsenic (As) | ≤1.5 ppm |
Heavy Metals (Pb) | ≤1.5 ppm |
Lead (Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...