Flavin adenine dinucleotide disodium salt ≥95% (dry basis)
Supplier: Thermo Fisher Scientific
J15785-03
J15785-ME
101170-878EA
342.69
USD
101170-878
101169-864
Flavin adenine dinucleotide disodium salt ≥95% (dry basis)
Flavin adenine dinucleotide disodium salt
Contains a varying amount of moisture ranging from a dihydrate to a tetrahydrate at the time of packaging.
Formula:
C₂₇H₃₃N₉O₁₅P₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00151217 CAS Number: 84366-81-4 EINECS: 282-733-8 |
Specification Test Results
Assay (UV/vis - (450 nm, pH 7.0) | ≥95.0% (dry basis, disodium salt) |
Water Content (Karl Fischer |
≤8.0% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...