EDTA tetrasodium salt dihydrate 99-101%, Ultrapure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethylenedinitrilotetra-acetic acid tetrasodium salt dihydrateTetrasodium Ethylenediaminetetraacetate DihydrateEdetic acid tetrasodium salt dihydrateEthylenediaminetetra-acetic acid tetrasodium salt dihydrateEthylenediaminetetraacetic acid sodium hydrateKestranal® 4S dihydrateTetrasodium ethylenediaminetetraacetate dihydrateKestranal® 4S 2 H2O

J15700-A1 J15700-C4
103728-092EA 1865.49 USD
103728-092 101175-024
EDTA tetrasodium salt dihydrate 99-101%, Ultrapure
EDTA tetrasodium salt dihydrate
Formula: C₁₀H₁₂N₂Na₄O₈·2H₂O
MW: 416.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150026
CAS Number: 10378-23-1

Order Now

Specification Test Results

Form White crystalline powder
Assay (unspecified) 99 to 101% (as is)
Identification (FTIR) Conforms
pH (1%, H2O) 10.7 to 11.7
pH (5%, H2O) 10.7 to 12.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR