Bottle Top Dispensers, Sartorius
Supplier: Sartorius
LH-723074 LH-723075 LH-723071 LH-723063 LH-723061 LH-723065 LH-723070 LH-723070 LH-723075 LH-723062 LH-723061 LH-723064 LH-723071 LH-723074 LH-723073 LH-723073 LH-723060 LH-723065 LH-723060 LH-723064 LH-723072 LH-723072 LH-723062 LH-723063
75995-130EA 757.62 USD
CA75995-130 75995-130 75995-132 CA75995-134 75995-134 75995-126 CA75995-132 75995-136 CA75994-988 75994-988 75994-986 CA75994-986 75994-992 CA75994-992 75994-990 CA75994-994 75994-994 CA75994-990 75995-128 CA75995-138 75995-138 CA75995-128 CA75995-136 CA75995-126
Bottle Top Dispensers, Sartorius
Bottle Top Dispensers
Prospenser and Prospenser Plus are high quality bottle-top dispensers that offer excellent chemical resistance.
  • Incremental volume setup ensures accurate and repeatable results
  • Fully autoclavable
  • Smooth and light to operate
  • Easy to attach aspiration tube with a secure threaded connection
  • User-replaceable valves and wide range of accessories are available
Adavanced features of Prospenser Plus include, a media recirculation system that allows unused media to be recovered from the device and returned to the bottle, 360° rotating dispensing head, and a universal connector at the back of the unit for drying tubes and filters.
These dispensers are safe, convenient, and customizable, making them perfect for a wide range of dispensing needs. Extremely high chemical and thermal resistance due to ceramic piston, glass cylinder, and highly resistant fluoroplastic parts.
Información para pedidos: Bottle not included.
Información de suministro: All Prospenser Plus models have A45 thread as a default and they are supplied with A32, A38, and S40 bottle adapters. All Prospenser models have A32 thread as a default and they are supplied with A28, S40, and A45 bottle adapters.