Human Recombinant Growth and differentiation factor-5
Supplier: Enzo Life Sciences
201-802-0010 201-802-0050
76045-520EA 206.01 USD
76045-520 76045-522 76288-318
Human Recombinant Growth and differentiation factor-5
Proteins and Peptides
Produced in E. coli. Non-glycosylated homodimer, containing two 117 amino acids chains. To enable bacterial expression the N-terminal sequence of Ala-Pro-Leu-Thr was replaced with a Lys.
  • High purity
  • Carrier-free