RNA sodium salt (from yeast)
Supplier: Thermo Fisher Scientific
J21190-A1
J21190-22
J21190-14
J21190-A7
101173-886EA
640.52
USD
101173-886
101171-144
101171-806
103728-108
RNA sodium salt (from yeast)
RNA sodium salt
Storage Temperature:
Ambient |
CAS Number:
73049-77-1 EINECS: 277-256-7 |
Specification Test Results
Form | White powder |
Assay (unspecified) | Biuret test Negative |
Residual water | 4.0 - 8.0% |
Impurity content: Nitrogen | ≥14.5% |
Impurity content: Phosphorous | ≥ 8.0% |
pH (1% H2O) | 6.5 ± 0.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...