Anti-ZKSCAN1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20958
102712-786EA 536.92 USD
102712-786
Anti-ZKSCAN1 Rabbit Polyclonal Antibody
Antibodies
The ZKSCAN1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ZKSCAN1. This antibody reacts with human. The ZKSCAN1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: ZKSCAN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR