Anti-Recombinant Monoclonal Antibody [clone: Mouse Fc G2a]

Supplier: Bio X Cell
BE0097-50MG BE0097-1MG BE0097-100MG BE0097-5MG BE0097-25MG
103252-646EA 3920 USD
103252-646 103252-648 103252-650 103252-652 103572-512 76554-346 76554-704 76555-158 76556-210 76556-276
Anti-Recombinant Monoclonal Antibody [clone: Mouse Fc G2a]
Antibodies
InVivoMab recombinant mouse IgG2a Fc, Clone: mouse Fc G2a, Isotype: Rabbit IgG, Applications: in vivo administration

Type: Primary
Antigen: Recombinant
Clonality: Monoclonal
Clone: Mouse Fc G2a
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR