AlphaTec® 58-735 Medium-Duty Chemical and Cut Protection Gloves, Ansell
Supplier: Ansell Healthcare
58735060 58735070 58735080 58735090 58735100 58735110
76170-080CS 1740.5 USD
76170-080 76170-082 76170-084 76170-268 76170-270 76170-272
AlphaTec® 58-735 Medium-Duty Chemical and Cut Protection Gloves, Ansell
Gloves
Advanced chemical protection with a great level of cut defense.
  • INTERCEPT™ cut resistant technology provides protection against lacerations
  • Optimized fit integrates the cut liner into a nitrile shell for better dexterity and tactility versus double gloving
  • Excellent dry and wet grip enhance dexterity and reduce hand fatigue
  • High-visibility cut liner acts as an indicator for when glove is cut as chemical protection is compromised
  • Cuff can be folded to act as a gutter to prevent chemicals running down the arm
  • Liner material: Ansell INTERCEPT™ Technology
Medium-duty nitrile chemical protection protects workers from select chemical risks, along with long-lasting snag, puncture, and abrasion resistance.
INTERCEPT™ cut resistance technology provides a great level of cut protection along with great comfort while maintaining high dexterity and ease of donning and removal. ANSELL GRIP™ Technology allows for less force and optimal control and confidence when gripping in oily or wet environments, further reducing hand fatigue and enhancing worker comfort.
Palm dipped for further enhanced flexibility.
Certificaciones: Gloves meet ANSI Abrasion Level 6 and ANSI Cut Level A3.