Anti-Glypican 5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-14968
102705-138EA 660 USD
102705-138
Anti-Glypican 5 Rabbit Polyclonal Antibody
Antibodies
The Glypican 5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Glypican 5. This antibody reacts with human. The Glypican 5 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Glypican 5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR