Anti-MKK6/MEK6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-17271
102709-340EA 660 USD
102709-340
Anti-MKK6/MEK6 Rabbit Polyclonal Antibody
Antibodies
The MKK6 / MEK6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to MKK6 / MEK6. This antibody reacts with human. The MKK6 / MEK6 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: MKK6/MEK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR