Anti-HOXC8 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-16875
102708-710EA 536.92 USD
102708-710
Anti-HOXC8 Rabbit Polyclonal Antibody
Antibodies
The HOXC8 Antibody from Novus Biologicals is a rabbit polyclonal antibody to HOXC8. This antibody reacts with human. The HOXC8 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: HOXC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR