Kinesis® TELOS® neo™ MicroPlate™ SPE 96-Well Microplates, Antylia Scientific
Supplier: Antyila Scientific
06474-02 06474-05 06474-06 06474-09 06474-13 06474-15 06474-18 06474-21
76183-256EA 1047.53 USD
76183-256 76183-258 76183-262 76183-264 76183-266 76183-270 76183-278 76183-282 76183-288 76183-294
Kinesis® TELOS® neo™ MicroPlate™ SPE 96-Well Microplates, Antylia Scientific
Microplates Solid Phase Extraction Plates (SPE)
Versatile 96-well format for small volume biological fluid samples.
  • 50 µL elution volume for minimal sample dilution and reduced evaporation time
  • Optimized well geometry for 5 and 10 mg sorbent masses
  • Use with vacuum or positive pressure
The internal diameter, frit design, and sorbent material of the TELOS MicroPlate 96-well plates allow for maximum recovery with minimal sample dilution and reduced evaporation time. A well packed with 5 mg sorbent allows analytes to be eluted in as little as 50 µL.
Populated plate comes with 96 wells already in place. Modular design lets you simply remove any unwanted wells and add sealing strips. Full or partially populated plates can be processed using either vacuum or positive pressure.