MES sodium salt 99%

Supplier: Thermo Scientific Chemicals

Synonyms: MES-Na

AC39735-0250 AC39735-1000
103466-930EA 47.85 USD
103466-930 103466-932 103466-934
MES sodium salt 99%
MES sodium salt
Formula: C₆H₁₂NNaO₄S
Storage Temperature: Ambient
MDL Number: MFCD00065473
CAS Number: 71119-23-8

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Solubility (10 % in water) Clear colorless
UV at 280 nm A: ≤0.03
Titration after Ion exchange ≥98.5 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR