Adenosine 5'-monophosphate sodium salt hydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate hydrate(x:1:x), Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate hydrate, 5'-Adenylic acid,sodium salt, hydrate (9CI)

AC44071-0010 AC44071-0050
103518-918EA 25.29 USD
103518-918 103518-920 103518-922
Adenosine 5'-monophosphate sodium salt hydrate 99%
Adenosine 5'-monophosphate sodium salt hydrate
Formula: C₁₀H₁₄N₅Na₂O₈P
MW: 409.200 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065023
CAS Number: 149022-20-8

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Water ≤20.00 %
HPLC ≥98.5 %
pH 4.50 to 6.50
Specific optical rotation -41° to -43° (20 °C, 589 nm) (c=2 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR