2,2':6',2''-Terpyridine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
TERPY
AC21563-0050
AC21563-2500
103526-744EA
132
USD
103526-744
103526-746
103526-748
2,2':6',2''-Terpyridine 96%
2,2':6',2''-Terpyridine
Formula:
C₁₅H₁₁N₃ MW: 233.27 g/mol Boiling Pt: 370 °C Melting Pt: 89…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006213 CAS Number: 1148-79-4 UN: 2811 ADR: 6.1,I |
Specification Test Results
Appearance | Cream to yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 89 to 95 °C |
GC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...