Sterile Stainless Steel Scalpels, Propper Manufacturing
Supplier: Propper
12901000 12901100 12901500 12902100 12902200
76044-668BX 32.14 USD
76044-668 76044-670 76044-672 76044-674 76044-676
Sterile Stainless Steel Scalpels, Propper Manufacturing
Scalpels and Scalpel Assemblies Scalpels
Propper sterile stainless steel scalpels provide a razor-sharp cutting edge for superior cutting and clean incisions.
  • Disposable, sterile stainless steel scalpel
  • Individually wrapped, peel-back blister pack
Sterile scalpels are individually wrapped in peel-back packaging for ease of use.
#10: Curved cutting edge is one of the more traditional blade shapes, making small incisions to the skin and muscle clean and precise.
#11: Elongated triangular blade with a strong tip, ideal for stab incisions. Most commonly used for the incisions of chest drains, and opening various coronary arteries.
#15: Small curved cutting edge is the most popular blade for short and precise incisions.
#21: A large version of the No.10 blade with a curved cutting edge and a flat, unsharpened back edge.
#22: A tad bigger than the number #21 blade, used for skin incisions in both cardiac and thoracic surgery.