Anti-BAF57 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15540
102706-182EA 660 USD
102706-182
Anti-BAF57 Rabbit Polyclonal Antibody
Antibodies
The BAF57 Antibody from Novus Biologicals is a rabbit polyclonal antibody to BAF57. This antibody reacts with human, mouse. The BAF57 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: BAF57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR