SqualVax, OZ Biosciences
Supplier: OZ Biosciences
SqualVax is an oil-in-water emulsion made of squalene droplets in a continuous aqueous phase similar to MF59®
Squalene emulsion induces local stimulation and recruitment of DCs and granulocytes, differentiation of monocytes into DCs and increased uptake of antigen by DCs. The emulsion acts more specifically on macrophages present at the site of injection. This formulation enhances differentiation of monocytes towards a mature phenotype, thereby promoting migration of antigen-loaded cells to the draining lymph node.
It is fully biodegradable, which is an important advantage over alternative oils that have been used in emulsion adjuvants, like Freund’s adjuvant that contains mineral oil (paraffin oil) and has long term persistence in the organisms.
Información de suministro: Shipped at room temperature
Envase: Store at +4°C