Mag4C-LV Transfection Trial Kit, OZ Biosciences
Supplier: OZ Biosciences
Mag4C-LV is designed for magnetic virus concentration
- Simple, Rapid & Ready-to-use
- Concentration of lentiviruses by magnetic capture in 30-45 minutes
- High yield of viral capture and recovery
- No need to process magnetic beads before capture
- Avoid ultracentrifugation, precipitation and chemicals. Reduced handling steps of viruses
- Suitable for all conditions & viruses
- Improve virus stability under storage conditions
Mag4C-LV magnetic nanoparticles capture viruses in culture media with 80-99 % efficiency. Once captured onto magnetic beads, viruses can be:
Concentrated and stored with the Storage Buffer or directly used for downstream assay.
Concentrated, eluted from the magnetic beads with the Elution Buffer and stored with the Storage Buffer or used for subsequent assays.
This kit is specifically developed for capturing, concentrating and storing lentiviruses and retroviruses. This kit is composed of 3 reagents allowing Magnetic Capture/Concentration, Elution and Conservation of Lentiviruses and Retroviruses.
Información de suministro: Shipped at room temperature
Envase: Store at +4°C