PBS 10X, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
11PBS10X02 11PBS10X03
76180-740EA 173.79 USD
76180-740 76180-742
PBS 10X, MP Biomedicals
Buffers Electrophoresis Buffers Washing Buffers
PBS 10X is frequently used as a wash and dilution buffer.
  • Rnase, Dnase, Protease free
  • Filtered through 0,2µm PTFE
  • Suitable for cell culture
PBS 10 X buffer is ideal for maintaining a constant pH, and since it is isotonic and non-toxic to cells, it can be used in a wide variety of biological applications.
PBS is also used to take a reference spectrum when measuring protein adsorption.
Order Now
SPECIFICATIONS
Environmentally Preferable