HyFlex® 11-542 Lightest Weight High Cut Solutions Gloves, Ansell
Supplier: Ansell Healthcare
11542060 11542070 11542100 11542110
76020-730CS 3882.98 USD
76020-730 76020-732 76020-734 76020-736 76020-760 76020-762
HyFlex® 11-542 Lightest Weight High Cut Solutions Gloves, Ansell
Gloves
Extreme comfort and protection in high cut risk applications for increased compliance and an uninterrupted workday.
  • INTERCEPT™ Cut resistant technology platform
  • Lightest weight cut A7 delivering dexterity and comfort
  • On average lasts 2X longer than competitive set
  • Intermittent contact heat protection up to 212 °F
  • Fiberglass-free, NRL-free, and silicone-free
  • Liner material: Ansell INTERCEPT™ Technology
Exclusive dirt masking design and high abrasion resistance ensures a longer product life. Suitable for use across a wide range of applications and risks. Avoid the need to change gloves when transitioning between tasks.
Certificaciones: Meets ANSI Cut Level A7 and ANSI Abrasion Level 4.